The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010287.2 | NP_034417.1 | 3 - 4 | 930 | 82 |
GenBank mRNA | AK030359.1 | - | 1 - 2 | 1646 | 82 |
AK035356.1 | - | 3 - 4 | 975 | 82 | |
AK041527.1 | - | 2 - 3 | 820 | 82 | |
AK081276.1 | - | 3 - 4 | 943 | 82 | |
M80481.1 | - | 3 - 4 | 931 | 82 |