Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gsr
- Entrez Gene ID
-
14782
- Gene Name
- glutathione reductase
- Gene Aliases
-
AI325518, D8Ertd238e, Gr-1, Gr1
- Chromosome Location
-
Chr.8: 33653238 - 33698162 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X76341.2 |
- |
6 - 7 |
631 |
74 |
View Full Product Details