The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010398.3 | NP_034528.1 | 6 - 7 | 1064 | 93 |
GenBank mRNA | AB359230.2 | - | 6 - 7 | 1064 | 93 |
AF057279.2 | - | - | 1032 | 93 | |
AF082510.1 | - | 6 - 7 | 1032 | 93 | |
AK133502.1 | - | 6 - 7 | 1064 | 93 | |
M11284.1 | - | - | 1065 | 93 |