Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxd4
Entrez Gene ID
15436
Gene Name
homeobox D4
Gene Aliases
6030436D05Rik, Hox-4.2, Hox-5.1
Chromosome Location
Chr.2: 74721978 - 74729160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010469.2 NP_034599.2 3 - 4 322 75
GenBank mRNA AK136751.1 - - 239 75

View Full Product Details