The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008610.2 | NP_032636.1 | 10 - 11 | 1908 | 83 |
GenBank mRNA | AK148184.1 | - | 10 - 11 | 1907 | 83 |
AK163248.1 | - | 2 - 3 | 220 | 83 | |
AK179499.1 | - | 1 - 2 | 84 | ||
AK189970.1 | - | 1 - 2 | 108 | 83 | |
AK190237.1 | - | 1 - 2 | 108 | 83 | |
AK190797.1 | - | 1 - 2 | 108 | 83 | |
AK191078.1 | - | 1 - 2 | 110 | 84 | |
AK191300.1 | - | 1 - 2 | 108 | 83 | |
AK191758.1 | - | 1 - 2 | 108 | 83 | |
AK192637.1 | - | 1 - 2 | 108 | 83 | |
AK192709.1 | - | 1 - 2 | 108 | 83 | |
AK193954.1 | - | 1 - 2 | 108 | 83 | |
AK195150.1 | - | 1 - 2 | 108 | 83 | |
AK197653.1 | - | 1 - 2 | 108 | 83 | |
AK198474.1 | - | 1 - 2 | 108 | 83 | |
AK198639.1 | - | 1 - 2 | 108 | 83 | |
AK198965.1 | - | 1 - 2 | 109 | 83 | |
AK199060.1 | - | 1 - 2 | 108 | 83 | |
AK200339.1 | - | 1 - 2 | 108 | 83 | |
AK200683.1 | - | 1 - 2 | 108 | 83 | |
AK200774.1 | - | 1 - 2 | 83 | ||
AK201563.1 | - | 1 - 2 | 108 | 83 | |
AK202079.1 | - | 1 - 2 | 108 | 83 | |
AK202191.1 | - | 1 - 2 | 109 | 83 | |
AK203427.1 | - | 1 - 2 | 108 | 83 | |
AK203661.1 | - | 1 - 2 | 108 | 83 | |
AK203919.1 | - | 1 - 2 | 108 | 83 | |
AK204187.1 | - | 1 - 2 | 109 | 84 | |
AK204539.1 | - | 1 - 2 | 108 | 83 | |
AK204698.1 | - | 1 - 2 | 108 | 83 | |
AK204944.1 | - | 1 - 2 | 108 | 83 | |
AK219401.1 | - | 1 - 2 | 108 | 83 | |
BC070430.1 | - | 10 - 11 | 1908 | 83 | |
M84324.1 | - | 10 - 11 | 1654 | 83 |