Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp2
Entrez Gene ID
17390
Gene Name
matrix metallopeptidase 2
Gene Aliases
Clg4a, GelA, MMP-2
Chromosome Location
Chr.8: 92827292 - 92853420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK148184.1 - 11 - 12 2065 94
M84324.1 - 11 - 12 1812 94

View Full Product Details