The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289450.1 | NP_001276379.1 | 13 - 14 | 1013 | 124 |
NM_010597.4 | NP_034727.3 | 12 - 13 | 1429 | 124 | |
GenBank mRNA | AF033003.1 | - | 12 - 13 | 1784 | 124 |
AK004666.1 | - | 13 - 14 | 997 | 124 | |
AK015412.1 | - | 14 - 15 | 1269 | 124 | |
AK138467.1 | - | 12 - 13 | 1418 | 124 | |
AK153200.1 | - | 13 - 14 | 1015 | 124 | |
BC014701.1 | - | 12 - 13 | 1154 | 124 | |
U65591.1 | - | 12 - 13 | 1025 | 124 | |
X97281.1 | - | 12 - 13 | 1413 | 124 |