The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122899.1 | NP_001116371.1 | 9 - 10 | 1947 | 97 |
NM_010704.2 | NP_034834.1 | 9 - 10 | 1947 | 97 | |
NM_146146.2 | NP_666258.2 | 9 - 10 | 1947 | 97 | |
GenBank mRNA | AK140785.1 | - | 9 - 10 | 1449 | 97 |
AK143989.1 | - | 9 - 10 | 1636 | 97 | |
AK155891.1 | - | 9 - 10 | 1487 | 97 | |
BC082551.1 | - | 9 - 10 | 1432 | 97 | |
U42467.1 | - | 9 - 10 | 1455 | 97 | |
U46135.1 | - | 9 - 10 | 1455 | 97 | |
U49106.1 | - | 8 - 9 | 1395 | 97 | |
U49109.1 | - | 8 - 9 | 1395 | 97 | |
U49110.1 | - | 8 - 9 | 1395 | 97 | |
U52915.1 | - | 9 - 10 | 1909 | 97 | |
U58861.1 | - | 9 - 10 | 1477 | 97 | |
U58862.1 | - | 9 - 10 | 1487 | 97 | |
U58863.1 | - | 9 - 10 | 1477 | 97 | |
Y10296.1 | - | 8 - 9 | 1403 | 97 | |
Y10298.1 | - | 8 - 9 | 1403 | 97 |