The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199068.2 | NP_951031.2 | 1 - 2 | 546 | 74 |
GenBank mRNA | AK144418.1 | - | 1 - 2 | 439 | 74 |
AK147375.1 | - | 1 - 2 | 544 | 74 | |
BC060238.1 | - | 1 - 2 | 526 | 74 | |
L26507.1 | - | 1 - 2 | 522 | 74 | |
U95016.1 | - | 1 - 2 | 522 | 74 |