Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myh6
Entrez Gene ID
17888
Gene Name
myosin, heavy polypeptide 6, cardiac muscle, alpha
Gene Aliases
A830009F23Rik, AA517445, Myhc-a, Myhca, alpha-MHC
Chromosome Location
Chr.14: 54941921 - 54968014 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164171.1 NP_001157643.1 8 - 9 940 86
NM_010856.4 NP_034986.1 8 - 9 876 86
GenBank mRNA AK142232.1 - 8 - 9 913 86
BC061145.1 - 8 - 9 909 86
BC090979.1 - 8 - 9 909 86
BC110700.1 - 8 - 9 922 86
M76599.1 - 6 - 7 738 86
M76600.1 - 6 - 7 738 86
M76601.1 - 8 - 9 840 86

View Full Product Details