The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164171.1 | NP_001157643.1 | 15 - 16 | 2090 | 70 |
NM_010856.4 | NP_034986.1 | 15 - 16 | 2026 | 70 | |
GenBank mRNA | AK142232.1 | - | 15 - 16 | 2063 | 70 |
BC061145.1 | - | 15 - 16 | 2059 | 70 | |
BC090979.1 | - | 15 - 16 | 2059 | 70 | |
BC110700.1 | - | 15 - 16 | 2072 | 70 | |
BC138027.1 | - | 14 - 15 | 1965 | 70 | |
M76598.1 | - | 13 - 14 | 1888 | 70 | |
M76599.1 | - | 13 - 14 | 1888 | 70 | |
M76600.1 | - | 13 - 14 | 1888 | 70 | |
M76601.1 | - | 15 - 16 | 1990 | 70 |