Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myl2
Entrez Gene ID
17906
Gene Name
myosin, light polypeptide 2, regulatory, cardiac, slow
Gene Aliases
MLC-2, MLC-2v, Mlc2v, Mylpc
Chromosome Location
Chr.5: 122100904 - 122113396 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010861.3 NP_034991.3 6 - 7 453 112
GenBank mRNA AK002367.1 - 6 - 7 454 112
AK146674.1 - 6 - 7 457 112
M91602.1 - 6 - 7 445 112

View Full Product Details