Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nnat
Entrez Gene ID
18111
Gene Name
neuronatin
Gene Aliases
5730414I02Rik, AW107673, Peg5
Chromosome Location
Chr.2: 157560078 - 157562525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291129.1 NP_001278058.1 1 - 2 182 73
NM_010923.3 NP_035053.1 1 - 2 182 73
GenBank mRNA AK003004.1 - 1 - 2 150 73
AK160626.1 - 1 - 2 150 73
BC036984.1 - 1 - 2 140 73
DV062509.1 - 1 - 2 204 73
X83570.1 - 1 - 2 137 73

View Full Product Details