Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nos2
Entrez Gene ID
18126
Gene Name
nitric oxide synthase 2, inducible
Gene Aliases
NOS-II, Nos-2, Nos2a, i-NOS, iNOS
Chromosome Location
Chr.11: 78920859 - 78960232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010927.3 NP_035057.1 22 - 23 2964 69
GenBank mRNA AF065919.1 - 22 - 23 2961 69
AF065920.1 - 22 - 23 2961 69
AF065921.2 - 22 - 23 2961 69
AF065922.2 - 21 - 22 2834 69
AF065923.2 - 21 - 22 2834 69
AY090567.1 - 21 - 22 2784 69
M84373.1 - 22 - 23 2965 69
M87039.1 - 22 - 23 3036 69
M92649.1 - 22 - 23 2972 69
U43428.1 - 22 - 23 2852 69

View Full Product Details