Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pax8
Entrez Gene ID
18510
Gene Name
paired box 8
Gene Aliases
Pax-8
Chromosome Location
Chr.2: 24420551 - 24480092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011040.4 NP_035170.1 11 - 12 1497 65
GenBank mRNA AK050717.1 - 11 - 12 1473 65
AK052610.1 - 11 - 12 1474 65
BC020526.1 - 11 - 12 1464 65
X57487.1 - 11 - 12 1454 65

View Full Product Details