The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011101.3 | NP_035231.2 | 16 - 17 | 2077 | 75 |
GenBank mRNA | AK147084.1 | - | 16 - 17 | 2078 | 75 |
AK163950.1 | - | 6 - 7 | 765 | 75 | |
BC096493.1 | - | 16 - 17 | 1871 | 75 | |
M25811.1 | - | 16 - 17 | 2111 | 75 | |
X52684.1 | - | 17 - 18 | 2018 | 75 | |
X52685.1 | - | 17 - 18 | 2018 | 75 |