Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkcg
Entrez Gene ID
18752
Gene Name
protein kinase C, gamma
Gene Aliases
PKCgamma, Pkcc, Prkcc
Chromosome Location
Chr.7: 3303532 - 3331099 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291434.1 NP_001278363.1 5 - 6 788 84
NM_011102.4 NP_035232.1 5 - 6 788 84
GenBank mRNA AK144419.1 - 5 - 6 790 84
BC111807.1 - 5 - 6 675 84
L28035.1 - 5 - 6 866 84
X67129.1 - 5 - 6 597 84

View Full Product Details