Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkce
Entrez Gene ID
18754
Gene Name
protein kinase C, epsilon
Gene Aliases
5830406C15Rik, PKC[e], PKCepsilon, Pkce, R75156
Chromosome Location
Chr.17: 86166971 - 86657919 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011104.3 NP_035234.1 14 - 15 2872 127
GenBank mRNA AF028009.1 - 14 - 15 2073 127
AF325507.1 - 14 - 15 2064 127
AK085351.1 - 15 - 16 2285 127

View Full Product Details