Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpinb2
Entrez Gene ID
18788
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 2
Gene Aliases
PAI-2, Planh2, ovalbumin
Chromosome Location
Chr.1: 107500892 - 107525600 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174170.1 NP_001167641.1 5 - 6 607 120
NM_011111.4 NP_035241.1 5 - 6 626 120
GenBank mRNA AK081487.1 - 5 - 6 608 120
AK090049.1 - 5 - 6 595 120
BC138815.1 - 4 - 5 635 120
BC138817.1 - 4 - 5 635 120
X16490.1 - 4 - 5 554 120

View Full Product Details