The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011146.3 | NP_035276.2 | 6 - 7 | 1317 | 105 |
GenBank mRNA | AB644275.1 | - | 7 - 8 | 1323 | 105 |
AY208183.1 | - | 6 - 7 | 1317 | 105 | |
AY208184.1 | - | 6 - 7 | 1317 | 105 | |
AY243585.1 | - | 6 - 7 | 1317 | 105 | |
BC021798.1 | - | 6 - 7 | 1317 | 105 | |
CT010340.1 | - | 6 - 7 | 1272 | 105 | |
EF062476.1 | - | 6 - 7 | 1308 | 105 | |
U01664.1 | - | 7 - 8 | 1349 | 105 | |
U01841.1 | - | 7 - 8 | 1672 | 105 | |
U09138.1 | - | 6 - 7 | 1311 | 105 | |
U10374.1 | - | 5 - 6 | 1182 | 105 |