The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011196.2 | NP_035326.2 | 2 - 3 | 1129 | 89 |
GenBank mRNA | BC058742.1 | - | 2 - 3 | 1129 | 89 |
D13321.1 | - | 2 - 3 | 1118 | 89 |