The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272023.1 | NP_001258952.1 | 4 - 5 | 878 | 83 |
NM_011325.2 | NP_035455.1 | 4 - 5 | 875 | 83 | |
NR_073548.1 | 4 - 5 | 875 | 83 | ||
GenBank mRNA | AF112186.1 | - | 4 - 5 | 802 | 83 |
AK028909.1 | - | 4 - 5 | 857 | 83 | |
AK036819.1 | - | 4 - 5 | 876 | 83 | |
AK087055.1 | - | 4 - 5 | 837 | 83 | |
AK164709.1 | - | 4 - 5 | 838 | 83 | |
BC131969.1 | - | 4 - 5 | 791 | 83 | |
BC132397.1 | - | 4 - 5 | 791 | 83 |