Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ccl27a
- Entrez Gene ID
-
20301
- Gene Name
- chemokine (C-C motif) ligand 27A
- Gene Aliases
-
ALP, AW558992, CTACK, CTAK, Ccl27, ESkine, ILC, PESKY, Scya27, Scya27a
- Chromosome Location
-
Chr.4: 41769467 - 41774210 on Build GRCm38
- Gene Symbol
- Ccl27b
- Entrez Gene ID
-
100040048
- Gene Name
- chemokine (C-C motif) ligand 27b
- Gene Aliases
-
Ctack, ENSMUSG00000066202, Scya27b
- Chromosome Location
-
Chr.4: 42650604 - 42656005 on Build GRCm38
- Gene Symbol
- Gm13306
- Entrez Gene ID
-
100039863
- Gene Name
- predicted gene 13306
- Gene Aliases
-
OTTMUSG00000011352
- Chromosome Location
-
Chr.4: 42153445 - 42158846 on Build GRCm38
- Gene Symbol
- Gm2506
- Entrez Gene ID
-
100039939
- Gene Name
- predicted gene 2506
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4|NT_187054: 7 - 3814 on Build GRCm38
- Gene Symbol
- LOC100861978
- Entrez Gene ID
-
100861978
- Gene Name
- c-C motif chemokine 27-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4|NT_187053: 16836 - 22237 on Build GRCm38
View Full Product Details