Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sele
Entrez Gene ID
20339
Gene Name
selectin, endothelial cell
Gene Aliases
CD62E, E-selectin, Elam
Chromosome Location
Chr.1: 164039604 - 164058487 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011345.2 NP_035475.1 4 - 5 662 110
GenBank mRNA AK153623.1 - 4 - 5 681 110
BC119083.1 - 4 - 5 613 110
BC120795.1 - 4 - 5 613 110
M87862.1 - 4 - 5 662 110

View Full Product Details