Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sell
Entrez Gene ID
20343
Gene Name
selectin, lymphocyte
Gene Aliases
AI528707, CD62L, L-selectin, LECAM-1, Lnhr, Ly-22, Ly-m22, Lyam-1, Lyam1
Chromosome Location
Chr.1: 164061813 - 164080785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011346.2 NP_035476.1 4 - 5 695 75
GenBank mRNA AB353767.1 - 3 - 4 582 75
AB353768.1 - 3 - 4 582 75
AK137509.1 - 4 - 5 607 75
AK157273.1 - 4 - 5 684 75
AK161754.1 - 4 - 5 75
AK163066.1 - 2 - 3 152 75
AK172116.1 - 4 - 5 685 75
BC052681.1 - 4 - 5 679 75
M25324.1 - 4 - 5 687 75
M36005.1 - 4 - 5 633 75
M36058.1 - 4 - 5 633 75
X14772.1 - 4 - 5 636 75

View Full Product Details