The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011348.2 | NP_035478.2 | 4 - 5 | 1093 | 69 |
GenBank mRNA | AF034744.1 | - | 4 - 5 | 828 | 69 |
AK141295.1 | - | 4 - 5 | 1095 | 69 | |
AK142340.1 | - | 4 - 5 | 920 | 69 | |
AK159214.1 | - | 4 - 5 | 884 | 69 | |
BC057956.1 | - | 4 - 5 | 991 | 69 | |
Z80941.1 | - | 4 - 5 | 459 | 69 | |
Z93947.1 | - | 4 - 5 | 1068 | 69 | |
Z93948.1 | - | 4 - 5 | 1068 | 69 |