Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema6c
Entrez Gene ID
20360
Gene Name
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C
Gene Aliases
Semay, mKIAA1869
Chromosome Location
Chr.3: 95160420 - 95174050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272024.1 NP_001258953.1 12 - 13 1459 79
NM_011351.2 NP_035481.1 9 - 10 1027 79
NR_073549.1 10 - 11 1352 79
NR_073550.1 10 - 11 1366 79
NR_073551.1 11 - 12 1415 79
GenBank mRNA AB013729.1 - 9 - 10 992 79
AF363972.1 - 8 - 9 940 79
AF363973.1 - 9 - 10 1060 79
AK031131.1 - 10 - 11 1264 79
AK129459.1 - 4 - 5 503 79
AK161042.1 - 4 - 5 506 79
AK165085.1 - 10 - 11 1279 79
BC051979.1 - 11 - 12 1321 79
BC059007.1 - 12 - 13 1371 79

View Full Product Details