The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164480.1 | NP_001157952.1 | 6 - 7 | 1420 | 58 |
NM_001164481.1 | NP_001157953.1 | 7 - 8 | 1674 | 58 | |
NM_001164482.1 | NP_001157954.1 | 5 - 6 | 1359 | 58 | |
NM_001164568.1 | NP_001158040.1 | 5 - 6 | 1389 | 58 | |
NM_011379.4 | NP_035509.4 | 6 - 7 | 1643 | 58 | |
GenBank mRNA | AK028151.1 | - | 5 - 6 | 630 | 58 |
AK137578.1 | - | 6 - 7 | 1366 | 58 | |
AK149669.1 | - | 6 - 7 | 1366 | 58 | |
AK156280.1 | - | 7 - 8 | 1628 | 58 | |
AK169945.1 | - | 5 - 6 | 1305 | 58 | |
AK170246.1 | - | 5 - 6 | 1319 | 58 | |
AK220472.1 | - | 6 - 7 | 1556 | 58 | |
BC054824.1 | - | 6 - 7 | 1364 | 58 | |
D11374.1 | - | 5 - 6 | 1317 | 58 |