The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164480.1 | NP_001157952.1 | 8 - 9 | 1672 | 92 |
NM_001164481.1 | NP_001157953.1 | 9 - 10 | 1926 | 92 | |
NM_001164482.1 | NP_001157954.1 | 7 - 8 | 1611 | 92 | |
NM_001164568.1 | NP_001158040.1 | 7 - 8 | 1641 | 92 | |
NM_011379.4 | NP_035509.4 | 8 - 9 | 1895 | 92 | |
GenBank mRNA | AK028151.1 | - | 7 - 8 | 882 | 92 |
AK137578.1 | - | 8 - 9 | 1618 | 92 | |
AK149669.1 | - | 8 - 9 | 1618 | 92 | |
AK156280.1 | - | 9 - 10 | 1880 | 92 | |
AK169945.1 | - | 7 - 8 | 1557 | 92 | |
AK170246.1 | - | 7 - 8 | 1571 | 92 | |
AK220472.1 | - | 8 - 9 | 1808 | 92 | |
BC054824.1 | - | 8 - 9 | 1616 | 92 | |
D11374.1 | - | 7 - 8 | 1569 | 92 |