The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079690.1 | NP_001073158.1 | 16 - 17 | 2268 | 71 |
NM_183354.2 | NP_899197.2 | 16 - 17 | 2268 | 71 | |
GenBank mRNA | BC016888.1 | - | 15 - 16 | 2246 | 71 |
BC051100.1 | - | 9 - 10 | 1032 | 71 | |
U20973.1 | - | 16 - 17 | 2266 | 71 | |
U20974.1 | - | 16 - 17 | 2266 | 71 | |
U20975.1 | - | 16 - 17 | 2266 | 71 | |
U61381.1 | - | 15 - 16 | 2237 | 71 | |
U94518.1 | - | 15 - 16 | 2215 | 71 |