Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tagln
Entrez Gene ID
21345
Gene Name
transgelin
Gene Aliases
Sm22, Sm22a, Ws310
Chromosome Location
Chr.9: 45929628 - 45936058 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011526.5 NP_035656.1 3 - 4 596 83
GenBank mRNA AK002880.1 - 3 - 4 435 83
AK077235.1 - 3 - 4 437 83
BC003795.1 - 3 - 4 425 83
L41154.1 - 3 - 4 435 83
U36588.1 - 3 - 4 435 83

View Full Product Details