The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011611.2 | NP_035741.2 | 1 - 2 | 121 | 68 |
NM_170702.2 | NP_733803.2 | 1 - 2 | 121 | 68 | |
NM_170703.2 | NP_733804.1 | 1 - 2 | 121 | 68 | |
NM_170704.2 | NP_733805.1 | 1 - 2 | 121 | 68 | |
NR_027852.1 | 1 - 2 | 121 | 68 | ||
GenBank mRNA | AJ401387.1 | - | 1 - 2 | 55 | 68 |
AJ401388.1 | - | 1 - 2 | 55 | 68 | |
AJ401389.1 | - | 1 - 2 | 55 | 68 | |
AJ401390.1 | - | 1 - 2 | 55 | 68 | |
AK089861.1 | - | - | 74 | 68 | |
AK150959.1 | - | 1 - 2 | 78 | 68 | |
AK151458.1 | - | 1 - 2 | 78 | 68 | |
AK152716.1 | - | 1 - 2 | 68 | ||
AK152756.1 | - | 1 - 2 | 77 | 68 | |
AK152942.1 | - | 1 - 2 | 85 | 68 | |
AK156644.1 | - | 1 - 2 | 89 | 68 | |
AK161978.1 | - | 1 - 2 | 78 | 68 | |
AK162305.1 | - | 1 - 2 | 131 | 68 | |
BY259264.1 | - | 1 - 2 | 118 | 68 | |
BY743050.1 | - | 1 - 2 | 77 | 68 | |
M83312.1 | - | 1 - 2 | 57 | 68 |