The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011611.2 | NP_035741.2 | 5 - 6 | 572 | 127 |
NM_170704.2 | NP_733805.1 | 5 - 6 | 572 | 127 | |
NR_027852.1 | 5 - 6 | 572 | 127 | ||
GenBank mRNA | AJ401388.1 | - | 5 - 6 | 506 | 127 |
AJ401389.1 | - | 5 - 6 | 506 | 127 | |
AK089861.1 | - | - | 525 | 127 | |
AK150959.1 | - | 5 - 6 | 529 | 127 | |
AK151458.1 | - | 5 - 6 | 529 | 127 | |
AK152716.1 | - | 5 - 6 | 530 | 127 | |
AK152756.1 | - | 5 - 6 | 528 | 127 | |
AK152942.1 | - | 5 - 6 | 536 | 127 | |
AK156644.1 | - | 5 - 6 | 540 | 127 | |
AK161978.1 | - | 5 - 6 | 529 | 127 | |
AK162305.1 | - | 5 - 6 | 582 | 127 | |
BC029254.1 | - | 5 - 6 | 530 | 127 | |
BY743050.1 | - | 5 - 6 | 528 | 127 | |
CT010343.1 | - | 5 - 6 | 500 | 127 | |
M83312.1 | - | 5 - 6 | 508 | 127 |