The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011773.3 | NP_035903.2 | 1 - 2 | 260 | 120 |
GenBank mRNA | AK144568.1 | - | 1 - 2 | 244 | 120 |
AK165796.1 | - | 1 - 2 | 223 | 120 | |
BC066199.1 | - | 1 - 2 | 242 | 120 | |
BY718969.1 | - | 1 - 2 | 263 | 120 | |
U76007.1 | - | 1 - 2 | 218 | 120 |