The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011775.6 | NP_035905.1 | 14 - 15 | 1703 | 94 |
GenBank mRNA | AK135903.1 | - | 14 - 15 | 1703 | 94 |
AK138699.1 | - | 12 - 13 | 1889 | 94 | |
BC071183.1 | - | 14 - 15 | 1685 | 94 | |
M34148.1 | - | 14 - 15 | 1703 | 94 |