Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Insrr
Entrez Gene ID
23920
Gene Name
insulin receptor-related receptor
Gene Aliases
Irr
Chromosome Location
Chr.3: 87796908 - 87816101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011832.2 NP_035962.2 13 - 14 3050 93
GenBank mRNA AB007135.1 - 13 - 14 2939 93
BC137855.1 - 13 - 14 2907 93

View Full Product Details