The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159500.1 | NP_001152972.1 | 5 - 6 | 1223 | 55 |
NM_011934.4 | NP_036064.3 | 5 - 6 | 1122 | 55 | |
GenBank mRNA | AK032025.1 | - | 5 - 6 | 930 | 55 |
AK044339.1 | - | 5 - 6 | 1123 | 55 | |
AK052256.1 | - | 5 - 6 | 1225 | 55 | |
BC044858.1 | - | 4 - 5 | 792 | 55 | |
BC132595.1 | - | 5 - 6 | 1087 | 55 | |
BC132597.1 | - | 5 - 6 | 1088 | 55 | |
S82458.1 | - | 4 - 5 | 830 | 55 | |
X89594.1 | - | 4 - 5 | 830 | 55 |