Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grk6
Entrez Gene ID
26385
Gene Name
G protein-coupled receptor kinase 6
Gene Aliases
Gprk6
Chromosome Location
Chr.13: 55445072 - 55460927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038018.4 NP_001033107.1 2 - 3 328 64
NM_001112711.2 NP_001106182.1 2 - 3 328 64
NM_001286063.1 NP_001272992.1 2 - 3 308 64
NM_001286064.1 NP_001272993.1 2 - 3 328 64
NM_001286065.1 NP_001272994.1 2 - 3 328 64
NM_001286066.1 NP_001272995.1 2 - 3 328 64
NM_011938.4 NP_036068.2 2 - 3 328 64
GenBank mRNA AF040747.1 - 2 - 3 198 64
AF040748.1 - 2 - 3 198 64
AF040749.1 - 2 - 3 198 64
AK051405.1 - 2 - 3 309 64
AK146119.1 - 2 - 3 203 64
BC057206.1 - 2 - 3 317 64
BC075719.1 - 2 - 3 209 64
CJ179645.1 - 2 - 3 329 64
Y15797.1 - 2 - 3 150 64
Y15798.1 - 2 - 3 172 64
Y15799.1 - 2 - 3 155 64
Y15800.1 - 2 - 3 150 64

View Full Product Details