The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038018.4 | NP_001033107.1 | 13 - 14 | 1583 | 100 |
NM_001112711.2 | NP_001106182.1 | 13 - 14 | 1583 | 100 | |
NM_001286063.1 | NP_001272992.1 | 13 - 14 | 1563 | 100 | |
NM_001286064.1 | NP_001272993.1 | 13 - 14 | 1568 | 100 | |
NM_001286065.1 | NP_001272994.1 | 13 - 14 | 1481 | 100 | |
NM_001286066.1 | NP_001272995.1 | 13 - 14 | 1576 | 100 | |
NM_011938.4 | NP_036068.2 | 13 - 14 | 1583 | 100 | |
GenBank mRNA | AF040747.1 | - | 13 - 14 | 1453 | 100 |
AF040748.1 | - | 13 - 14 | 1453 | 100 | |
AF040749.1 | - | 13 - 14 | 1453 | 100 | |
AK051405.1 | - | 13 - 14 | 1565 | 100 | |
AK146119.1 | - | 13 - 14 | 1356 | 100 | |
AK161142.1 | - | 11 - 12 | 1215 | 100 | |
AK180303.1 | - | 1 - 2 | 43 | 100 | |
BC057206.1 | - | 13 - 14 | 1557 | 100 | |
BC075719.1 | - | 13 - 14 | 1464 | 100 | |
CA528447.1 | - | 2 - 3 | 182 | 100 | |
Y15797.1 | - | 13 - 14 | 1405 | 100 | |
Y15798.1 | - | 13 - 14 | 1427 | 100 | |
Y15799.1 | - | 13 - 14 | 1410 | 100 | |
Y15800.1 | - | 13 - 14 | 1398 | 100 |