Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpc7
Entrez Gene ID
26946
Gene Name
transient receptor potential cation channel, subfamily C, member 7
Gene Aliases
TRP7, Trrp8
Chromosome Location
Chr.13: 56773098 - 56895974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF139923.1 - 10 - 11 2437 78
AK135153.1 - 10 - 11 2623 78
AK137418.1 - 11 - 12 2762 78
BC117995.1 - 10 - 11 2437 78

View Full Product Details