The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013477.3 | NP_038505.2 | 1 - 2 | 255 | 70 |
GenBank mRNA | AB088408.1 | - | 1 - 2 | 144 | 70 |
AK083372.1 | - | 1 - 2 | 256 | 70 | |
AK133921.1 | - | 1 - 2 | 263 | 70 | |
AK171515.1 | - | 1 - 2 | 225 | 70 | |
U13840.1 | - | 1 - 2 | 144 | 70 | |
U21549.1 | - | 1 - 2 | 127 | 70 |