Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp6v0d1
Entrez Gene ID
11972
Gene Name
ATPase, H+ transporting, lysosomal V0 subunit D1
Gene Aliases
AI267038, Ac39, Atp6d, P39, VATX, Vma6
Chromosome Location
Chr.8: 105524470 - 105566040 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013477.3 NP_038505.2 3 - 4 612 57
GenBank mRNA AB088408.1 - 3 - 4 501 57
AK083372.1 - 3 - 4 613 57
AK133921.1 - 3 - 4 620 57
AK171515.1 - 3 - 4 582 57
BC011075.1 - 3 - 4 503 57
U13840.1 - 3 - 4 501 57
U21549.1 - 3 - 4 484 57

View Full Product Details