The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013490.4 | NP_038518.2 | 2 - 3 | 753 | 73 |
GenBank mRNA | AB011002.1 | - | 2 - 3 | 612 | 73 |
BC060218.1 | - | 2 - 3 | 716 | 73 |