The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013520.3 | NP_038548.3 | 6 - 7 | 582 | 95 |
GenBank mRNA | CK622267.1 | - | 6 - 7 | 552 | 95 |
EU274583.1 | - | 5 - 6 | 494 | 95 | |
L23636.1 | - | 5 - 6 | 525 | 95 | |
U04807.1 | - | 5 - 6 | 749 | 95 |