The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161775.1 | NP_001155247.1 | 1 - 2 | 134 | 68 |
NM_013607.2 | NP_038635.2 | 1 - 2 | 134 | 68 | |
GenBank mRNA | AK085742.1 | - | 1 - 2 | 92 | 68 |
AK137226.1 | - | 1 - 2 | 91 | 68 | |
AK137716.1 | - | 1 - 2 | 90 | 68 | |
AK173045.1 | - | 1 - 2 | 98 | 68 | |
BC026142.1 | - | 1 - 2 | 119 | 68 | |
D85923.1 | - | 1 - 2 | 89 | 68 | |
D85924.1 | - | 1 - 2 | 89 | 68 | |
L25860.1 | - | 1 - 2 | 78 | 68 | |
S81516.1 | - | 1 - 2 | 61 | 68 |