Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh11
Entrez Gene ID
17880
Gene Name
myosin, heavy polypeptide 11, smooth muscle
Gene Aliases
AV071570, SM1, SM2, smMHC
Chromosome Location
Chr.16: 14194527 - 14291408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161775.1 NP_001155247.1 2 - 3 488 105
NM_013607.2 NP_038635.2 2 - 3 488 105
GenBank mRNA AK085742.1 - 2 - 3 446 105
AK137226.1 - 2 - 3 445 105
AK137716.1 - 2 - 3 444 105
AK173045.1 - 2 - 3 452 105
BC026142.1 - 2 - 3 473 105
D85923.1 - 2 - 3 443 105
D85924.1 - 2 - 3 443 105
L25860.1 - 2 - 3 432 105

View Full Product Details