Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh11
Entrez Gene ID
17880
Gene Name
myosin, heavy polypeptide 11, smooth muscle
Gene Aliases
AV071570, SM1, SM2, smMHC
Chromosome Location
Chr.16: 14194527 - 14291408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161775.1 NP_001155247.1 37 - 38 5446 71
NM_013607.2 NP_038635.2 37 - 38 5446 71
GenBank mRNA AK173045.1 - 37 - 38 5410 71
D85923.1 - 37 - 38 5401 71
D85924.1 - 37 - 38 5401 71

View Full Product Details