The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013652.2 | NP_038680.1 | 2 - 3 | 269 | 153 |
GenBank mRNA | AF128218.1 | - | 2 - 3 | 242 | 153 |
AF128219.1 | - | 2 - 3 | 242 | 153 | |
AK156948.1 | - | 2 - 3 | 262 | 153 | |
AK172472.1 | - | 2 - 3 | 262 | 153 | |
BC119257.1 | - | 2 - 3 | 216 | 153 | |
BC119259.1 | - | 2 - 3 | 216 | 153 | |
BY163868.1 | - | 2 - 3 | 270 | 153 | |
M23503.1 | - | 2 - 3 | 267 | 153 | |
M35590.1 | - | 2 - 3 | 252 | 153 |