Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema5b
Entrez Gene ID
20357
Gene Name
sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B
Gene Aliases
AI893641, SemG, Semag, mKIAA1445
Chromosome Location
Chr.16: 35541154 - 35664261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013661.2 NP_038689.2 4 - 5 585 55
GenBank mRNA AK140524.1 - 4 - 5 803 55
BC052397.1 - 4 - 5 585 55
X97818.1 - 4 - 5 376 55

View Full Product Details