Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema6b
Entrez Gene ID
20359
Gene Name
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B
Gene Aliases
Sema, Seman, VIb, semaZ
Chromosome Location
Chr.17: 56123085 - 56140343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130456.1 NP_001123928.1 11 - 12 1285 76
NM_013662.2 NP_038690.1 11 - 12 1396 76
GenBank mRNA AF036585.1 - 10 - 11 1152 76
AK139358.1 - 11 - 12 1205 76
AK143229.1 - 11 - 12 1286 76
AK166077.1 - 11 - 12 1388 76
BC139415.1 - 10 - 11 1151 76

View Full Product Details